Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

656054-1L
BJ656054-1LEA 58.02 USD InStock
BJ656054-1L
Sodium hydroxide 10.0 M NaOH (10.0N) volumetric solution, Fluka™
Sodium hydroxide
Honeywell's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
  • Masking agents
  • Indicators for volumetric and complexometric titrations

UNSPSC Code: 12000000


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 120 °C
Density: 1.350 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Assay 9.80 - 10.20 N
Carbonate (as Na₂CO₃) Max. 0.12 N
Traceable to NIST SRM effective

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty