Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum(VI) oxide ≥99.5%, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Molybdenum trioxide

69850-500G 69850-100G
BJ69850-100GEA 131.93 USD InStock
BJ69850-100G BJ69850-500G
Molybdenum(VI) oxide ≥99.5%, Puriss. p.a., Fluka™
Molybdenum (VI) oxide


Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1.155 °C
Melting Pt: 795 °C
Density: 4.70 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
Order Now
Specification Test Results
Assay Min. 99.8 %
Colour of the solution complying
Turbidity of solution Max. 5.5 NTU
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 10 ppm
Cobalt (Co) Max. 20 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 20 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 10 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 20 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 20 ppm
Chloride (Cl) Max. 20 ppm
Sulfate (SO₄) Max. 50 ppm
Ammonium (NH₄) Max. 50 ppm

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty