Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium borohydride Sodium tetrahydridoborate

71321-25G 71321-100G
BJ71321-25GEA 85.33 USD InStock
BJ71321-25G BJ71321-100G
Sodium borohydride ≥99%, TraceSELECT™ for the determination of hydride formers by aas, Fluka™
Sodium borohydride
TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Formula: NaBH₄
MW: 37.83 g/mol
Boiling Pt: 400 °C (1.027 hPa)
Melting Pt: 360 °C
Density: 1.074 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3,I
Order Now
Specification Test Results
Assay 98.5 - 101.5 %
Arsenic (As) Max. 0.05 ppm
Mercury (Hg) Max. 50 ppb
Antimony (Sb) Max. 0.05 ppm
Selenium (Se) Max. 0.05 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty