Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Sodium chloride ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

71380-1KG 71380-500G 71380-5KG
BJ71380-5KGEA 361.88 USD InStock
BJ71380-5KG BJ71380-500G BJ71380-1KG
Sodium chloride ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1.465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Order Now
Specification Test Results
Assay Min. 99.5 %
pH (5 %, 20°C) 5.0 - 8.0
Arsenic (As) Max. 0.00001 %
Barium (Ba) Max. 0.0005 %
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Cobalt (Co) Max. 0.0005 %
Chromium (Cr) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0001 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.0005 %
Manganese (Mn) Max. 0.0005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Bromide (Br) Max. 0.005 %
Iodide (I) Max. 0.001 %
Potassium hexacyanoferrate(II) Max. 0.0001 %
Phosphate (PO₄) Max. 0.0005 %
Sulfate (SO₄) Max. 0.001 %
Total N Max. 0.001 %
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty