Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ~50% in aqueous solution for HPLC, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

71686-1L 71686-2.5L
BJ71686-2.5LEA 217.67 USD InStock
BJ71686-2.5L BJ71686-1L
Sodium hydroxide ~50% in aqueous solution for HPLC, Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 143 °C
Melting Pt: 12 °C
Density: 1.525 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Assay 49.0 - 51.0 %
Density (D 20/4) 1.500 - 1.540
Carbonate (as Na₂CO₃) Max. 0.1 %
Absorbance 500 nm Max. 0.02
Absorbance at 300 nm Max. 0.03
Absorbance 280 nm Max. 0.08
Absorbance at 260 nm Max. 0.15
Absorbance at 240 nm Max. 0.17
Suitability test (HPLC) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty