Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 34 - 37%, TraceSELECT™ for inorganic trace analysis, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acidHydrochloric acid solution

72787-2.5L
BJ72787-2.5LEA 416.19 USD InStock
BJ72787-2.5L
Hydrochloric acid 34 - 37%, TraceSELECT™ for inorganic trace analysis, Fluka™
Hydrochloric acid
TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 34 - 37 %
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 5 ppb
Gold (Au) Max. 1 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 10 ppb
Cadmium (Cd) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Mercury (Hg) Max. 0.5 ppb
Indium (In) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 10 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 1 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 0.5 ppb
Free chlorine (Cl) Max. 0.5 ppm
Bromide (Br) Max. 10000 ppb
Phosphate (PO₄) Max. 50 ppb
Sulfate (SO₄) Max. 1000 ppb
Vapor Pressure 7.93 psi ( 37.7 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty