Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid fuming 37%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acid

84436-1L
BJ84436-1LEA 93.31 USD InStock
BJ84436-1L
Hydrochloric acid fuming 37%, tested according to Ph. Eur., Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Vapor Pressure 3.23 psi ( 21.1 °C)
Identity A complying
Identity B complying
Assay 35.0 - 39.0 %
Appearance of the solution complying
Non-volatile matter Max. 0.01 %
Sulfate (SO₄) Max. 20 ppm
Free chlorine (Cl) Max. 4 ppm
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty