Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

85228-250G 85228-100G 85228-50G
BJ85228-250GEA 1208.51 USD InStock
BJ85228-250G BJ85228-100G BJ85228-50G
Silver nitrate ≥99.5% (by argentometric titration), Puriss. p.a., Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 212 °C
Density: 4.350 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Assay 99.5 - 100.5 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 2 ppm
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 20 ppm

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty