Would you like to log in again?
Triethylammonium Acetate Buffer, BioUltra, Volatile Buffer, Fluka®, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
90357-100ML 90357-500ML
BJ90357-100MLEA 180.95 USD InStock
BJ90357-100ML BJ90357-500ML
Triethylammonium Acetate Buffer, BioUltra, Volatile Buffer, Fluka®, Honeywell Research Chemicals
Buffers Biological Buffers
  • Absorbance 260 nm: max. 0.03
  • Absorbance 280 nm: max. 0.03
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty