Synonyms:
Muriatic acid
, Hydrochloric acid solution
96208-500ML
96208-250ML
96208-1L
BJ96208-500MLEA
720.45
USD
InStock
BJ96208-500ML
BJ96208-250ML
BJ96208-1L
Hydrochloric acid 32 - 35%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Hydrochloric acid
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Beilstein Number: 1098214
RTECS Number: 1098214
UNSPSC Code: 12352301