Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Dichloromethane ≥99.9% stabilized, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with cyclohexene

BB301-4 BB301-4 CS301-56 BC301-200
BJBC301-200EA 4093.85 USD InStock
BJBC301-200 BJBB301-4 CABJBB301-4 BJCS301-56
Dichloromethane ≥99.9% stabilized, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Dichloromethane
BioSyn products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 40 °C
Melting Pt: -95 °C
Density: 1.33 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Acidity (as HCl) 1 mg/L max.
Chloride 10 mg/L max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.4236 - 1.4246
Residue 1 mg/L max.
UV Absorbance @ 240 nm 0.1 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Cutoff 233 nm max.
Water by Karl Fischer Titration 20 ppm max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty