Synonyms:
NSC 5356
, Formic acid dimethylamide
, Dimethylformamide
, DMFA
, DMF
, Formyldimethylamine
, N-Formyldimethylamine
BC075-200
BN075-20
BB075-4
BB075-4
BN075-20
BJBN075-20EA
2040.18
USD
InStock
BJBN075-20
BJBC075-200
BJBB075-4
CABJBB075-4
CABJBN075-20
N,N-Dimethylformamide ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
N,N-Dimethylformamide
BioSyn« products are ideal for DNA, RNA and oligonucleotide synthesis. All solvents and regents are filtered (0.2-micron) and tested for oxidation reage.
- Ultra-low water specifications
- Low amine specifications
- Low particulate levels
- High solvent purity