BR640-4X2.5L
BR640-4X4L
BR640-4X2.5L
BJBR640-4X2.5LCS
3874.38
USD
InStock
BJBR640-4X2.5L
CABJBR6404X2.5
BJBR640-4X4L
Capping Reagent 80/10/10 THF/Lutidine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination