Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capping Reagent 80/10/10 THF/Pyridine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

BR641-4X2.5L BR641-2 BR641-4 BR641-2 BR641-4X2.5L
BJBR641-4X2.5LCS 3207.88 USD InStock
CABJBR6414X2.5 BJBR641-4X2.5L CABJBR641-2 BJBR641-2 BJBR641-4
Capping Reagent 80/10/10 THF/Pyridine/Acetic Anhydride (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis.

  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


Boiling Pt: 66 °C (main component)
Melting Pt: -108.5 °C (main component)
Density: 0.888 g/cm³ (20 °C. The physical data is that of the main component)
Flash Pt: 11.8 %(V)
Storage Temperature: Ambient
UN: 2924
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty