Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

BR654-1 BR654-1
BJBR654-1CS 1269.62 USD InStock
CABJBR654-1 BJBR654-1
Capping Reagent 80/20 Acetonitrile/NMI (N-Methylimidazole) (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Capping Reagent
Reagents are specifically processed and purified to ensure the high synthesis efficiency

  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications

Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.


Boiling Pt: 86.6 °C (main component)
Melting Pt: -43.8 °C (main component)
Density: 0.827 g/cm³ (20 °C)
Flash Pt: 6 °C (main component)
Storage Temperature: Ambient
UN: 2924
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty