Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Oxidation reagent 0.02 M I₂ in 70/20/10 THF/pyridine/water (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

BR666-4X2.5L BR666-2 BR666-200 SR666-4 SR666-20 BR666-56 BR666-4X2.5L SR666-4
BJSR666-20EA 2618.27 USD InStock
BJSR666-20 BJBR666-2 BJSR666-4 BJBR666-200 CABJSR666-4 CABJBR66642.5L BJBR666-4X2.5L BJBR666-56
Oxidation reagent 0.02 M I₂ in 70/20/10 THF/pyridine/water (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Iodine
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

UNSPSC Code: 12350000


Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 66 °C
Melting Pt: -108.5 °C
Density: 0.935 g/cm3 (20 °C)
Flash Pt: -15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1993
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty