Synonyms:
DCA
BR676-4
BN676-20
BJBR676-4CS
1537.73
USD
InStock
BJBR676-4
BJBN676-20
Dichloroacetic acid (DCA) 10% in toluene, BioSyn™ Detritylation Reagent, for DNA/RNA synthesis, Burdick & Jackson™
Dichloroacetic acid (DCA)
Detritylation Solution contains 10% Dichloroacetic acid (DCA) in Toluene (v/v), suitable for use with DNA/RNA synthesis instruments. Packaged in 20-liter steel NOWPak containers that are returned for refill. Sold only under contract.