Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Chloro calcium

C1016-100G C1016-2.5KG C1016-500G
BJC1016-500GEA 106.06 USD InStock
BJC1016-500G BJC1016-2.5KG BJC1016-100G
Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1.900 °C (1.013 hPa)
Melting Pt: 765 °C
Density: 2.150 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 13,01660
Order Now
Specification Test Results
Form granular
Assay Min. 93.0 %
Colour of the solution complying
Clearness of the solution complying
Granulation complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty