Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Chloroform ≥99.5% stabilized

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene 100-200 ppm

C2432-500ML C2432-1L C2432-2.5L C2432-6X500ML C2432-4X25ML C2432-25ML
BJC2432-1LEA 143.52 USD InStock
BJC2432-1L BJC2432-4X25ML BJC2432-2.5L BJC2432-500ML BJC24326X500ML BJC2432-25ML
Chloroform ≥99.5% stabilized
Chloroform
We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Beilstein Number: 1731042
RTECS Number: FS9100000
UNSPSC Code: 12191502


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (1.013 hPa)
Melting Pt: -61.3 °C
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Vapor Pressure 160 mmHg ( 20 °C)
NMR-spectrum complying
Assay (GC) Min. 99.50 %
2-methyl-2-buten (GC) 20 - 200 ppm
Refractive Index n20/D 1.445 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty