Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

CS015-56 CS015-200
BJCS015-200EA 9286.25 USD InStock
BJCS015-200 BJCS015-56
Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™
Acetonitrile
  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
GC Analysis 99.9 % min.
Water by Karl Fischer Titration 0.005 % max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 1 mg/L max.
UV Absorbance @ 200 nm 0.05 AU max.
UV Absorbance @ 210 nm 0.04 AU max.
UV Absorbance @ 225 nm 0.01 AU max.
UV Absorbance @ 250 nm 0.005 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Cutoff 190 nm max.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty