Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Chloroform ≥99.9% stabilized, GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

GC049-4 GC049-4
BJGC049-4CS 2059.12 USD InStock
CABJGC049-4 BJGC049-4
Chloroform ≥99.9% stabilized, GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Chloroform
For trace analysis at or below the part per billion level. Ideal for EPA protocols.

  • Less than 1 ppb trace contamination for
  • Semi-volatiles with a molecular weight
  • Greater than toluene (92)
  • Ideal for EPA protocols
  • Actual lot chromatograms available upon request


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 62 °C
Melting Pt: -64 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Acidity (as HCl) 1 mg/L max.
Chloride 10 mg/L max.
Electron Capture GC (as lindane) 2 ppt max.
Flame Ionization GC (as nonane) 1 ppb max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.4454 - 1.4464
Residue 1 mg/L max.
Water by Karl Fisher Titration 0.02 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty