Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99.9% (by GC), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Pentane

GC312-4 GC312-4
BJGC312-4CS 1448.86 USD InStock
BJGC312-4 CABJGC312-4
n-Pentane ≥99.9% (by GC), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
n-Pentane
For trace analysis at or below the part per billion level (ppb). Ideal for EPA protocols.

  • Less than 1 ppb trace contamination for
  • Semi-volatiles with a molecular weight
  • Greater than toluene (92)
  • Ideal for EPA protocols
  • Actual lot chromatograms available upon request


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 7.8 %(V)
Melting Pt: -129.7 °C
Density: 0.626 g/cm³ (20 °C)
Flash Pt: -49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Electron Capture GC (as lindane) 2 ppt max.
Flame Ionization GC (as nonane) 1 ppb max.
GC Analysis (n-Pentane) 98 % min.
GC Analysis (total C5 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3573 - 1.3579
Residue 1 mg/L max.
Water by Karl Fischer Titration 0.01 % max.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty