Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

LP014TC19 LP014-4 LP014-4 LP014TC19 LP014-204 LP014-4P
BJLP014TC19EA 2323.93 USD InStock
CABJLP014TC19 BJLP014TC19 CABJLP014-4 BJLP014-4P BJLP014-4 BJLP014-204
Acetonitrile ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Acetonitrile
ACS general use certified, reagent grade

  • Low residue
  • Low water
  • High GC purity


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C
Melting Pt: -46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Appearance Passes test
Assay by GC Analysis 99.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.005 % max.
Tirable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
Water by Karl Fischer Titration 0.3 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty