Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Chloroform ≥99.8% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

LP049TC19 LP049TC19 LP049-4 LP049-4
BJLP049TC19EA 1972.71 USD InStock
BJLP049TC19 CABJLP049TC19 CABJLP049-4 BJLP049-4
Chloroform ≥99.8% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Chloroform
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 62 °C
Melting Pt: -64 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Appearance Passes test
Assay by GC Analysis 99.8 % min.
Color (APHA) 10 max.
Free Chlorine Passes test
Lead 0.05 ppm max.
Residue after Evaporation 0.001 % max.
Water by Karl Fischer Titration 0.02 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty