Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ethyl acetate ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: Honeywell Research Chemicals
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

LP100-4 LP100-4 LP100-4P LP100-19 LP100-19
BJLP100D19EA 987.99 USD InStock
CABJLP100-4 BJLP100D19 BJLP100-4 CABJLP100D19 BJLP100-4P
Ethyl acetate ≥99.5% ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Ethyl acetate
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C
Melting Pt: -84 °C
Density: 0.9006 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Assay by GC Analysis 99.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.003 % max.
Substances Darkened by Sulfuric Acid Passes test
Titrable Acid 0.9 µeq/g max.
Water by Karl Fischer Titration 0.2 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty