Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Diethyl ether ≥99.9% stabilized, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

PP108-19 PP108-19
BJPP108-19EA 1265.62 USD InStock
CABJPP108-19 BJPP108-19
Diethyl ether ≥99.9% stabilized, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Diethyl ether
For specialty organic synthesis and preparative LC. The 54-Gallon drum is individually analyzed with certificate on label. Stabilized with 1ppm BHT.

  • Low residue
  • Purified solvent
  • ACS General Use certified


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C
Melting Pt: -117.4 °C
Density: 0.713 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Alcohol (as CH3CH2OH) Passes test
Assay by GC Analysis 99.9 % min.
Carbonyl (as HCHO) 0.001 % max.
Color (APHA) 10 max.
Peroxides (as H₂O₂) at time of packaging 1 ppm max.
Refractive Index (20ºC) 1.3518 - 1.3538
Residue after Evaporation 5 mg/L max.
Titrable Acid 0.2 µeq/g max.
Water by Karl Fischer Titration 0.03 % max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty