Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Sodium chloride ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

S9888-500G S9888-2.5KG S9888-25KG S9888-6X500G S9888-4X2.5KG S9888-20KG S9888-1KG S9888-50KG S9888-10KG S9888-5KG
BJS9888-500GEA 76.31 USD InStock
BJS9888-500G BJS9888-1KG BJS9888-5KG BJS9888-25KG BJS9888-20KG BJS9888-4X2.5 BJS9888-10KG BJS9888-2.5KG BJS9888-6X500G BJS9888-50KG
Sodium chloride ≥99.0% ACS, Fluka™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1.465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Order Now
Specification Test Results
Assay Min. 99.0 %
pH (5 %, 25 °C) 5.0 - 9.0
Water insoluble matter Max. 0.005 %
Barium (Ba) complying
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (ICP) ACS Max. 5 ppm
Bromide (Br) Max. 0.01 %
Chlorate, Nitrate (as NO₃) Max. 0.003 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.004 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty