Would you like to log in again?
Multi-Disc™ Adapters for JS-4.0, JS-4.2, and JS-4.2A Rotors, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Multi-Disc™
339108 341977 341794 341794 339108 339101 339102 339102 339109 339101 339104 339100 345386 339109 345386 339100 339103
10769-310EA 284.63 USD InStock
CABK339109 10769-310 CABK339103 BK341977 CABK339101 CABK339102 CABK339108 BK341794 BK339100 BK339102 CABK345386 BK339101 BK339104 CABK339100 BK339108 BK339109 CABK341794
Multi-Disc™ Adapters for JS-4.0, JS-4.2, and JS-4.2A Rotors, Beckman Coulter®
Centrifuges Rotors
Suitable for use with the JS-4.0 (BK339086) and JS-4.2/JS-4.2A (BK339080 series) swinging bucket rotors.
  • For swinging bucket rotors
  • Color-coded
  • Stackable
Color-coded adapters consist of multiple discs that may be stacked to the desired tube or bottle length. Maximum length for multiple-place adapters is 130mm; maximum length for single-place adapters is 160mm. Adapters are autoclavable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty