Would you like to log in again?
JA-20.1 Fixed-Angle Rotor, Beckman Coulter®
Supplier: Beckman Coulter
342095 342095
BK342095EA 9524.06 USD InStock
CABK342095 BK342095
JA-20.1 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
This aluminum, 32-place, and fixed-angle rotor is for use with Avanti™ J-26 XP and XPIBK393124 series and J-EBK369001 and J6-MIBK360291 centrifuges.
  • Application: High-Force, Large-Volume Separation of Particles with 100S or larger sedimentation coefficients.
  • Material: Aluminum
  • Thirty-Two-Place Router
  • Fixed-Angle
  • Rotor Accommodates up to Thirty-Two 15 mL Tubes
It's ideal for high-force, large-volume separation of particles with 100S or larger sedimentation coefficients.
The rotor accommodates up to thirty-two 15 mL tubes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty