Would you like to log in again?
Tube Decanter for 10/12 mm Modular Disk Adapters, High Performance Rotors, Beckman Coulter®
Supplier: Beckman Coulter
343108 343108
BK343108EA 173.15 USD InStock
CABK343108 BK343108
Tube Decanter for 10/12 mm Modular Disk Adapters, High Performance Rotors, Beckman Coulter®
Centrifuges Rotors
For decanting a full rack of RIA tubes at once. Includes silicone gaskets for 10- and 12-mm tubes.
For use in blue modular disk adapters (part no. BK356148, BK359469).
For use in High Performance and High Capacity Rotors: JS-3.0 (BK339081) JS-5.2 (BK339087) JS-4.0 (BK339086) JS-4.2 (BK339080) JS-4.2A (BK366695).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty