Would you like to log in again?
JS-13.1 Swinging Bucket Rotor, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
346963 346963
BK346963EA 15253.28 USD InStock
BK346963 CABK346963
JS-13.1 Swinging Bucket Rotor, Beckman Coulter®
Centrifuges Rotors
This six-place, aluminum rotor with a swinging bucket is for use in Avanti™ J-26 XP and XPIBK393124 series and J6-MIBK360291 and J-EBK369001 centrifuges.
  • Applications: Harvesting Bacteria, Processing Tissue Homogenates, Separating Cell Particulates, and More
  • Space for Six 50 mL Tubes
  • Carrying Handle Included
  • Material: Aluminum
  • Swinging Bucket
It is ideal for harvesting bacteria, processing tissue homogenates, separating cell particulates, and isolating cell membrane using density gradients.
It accommodates six 50 mL tubes and includes a carrying handle.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty