Would you like to log in again?
Airfuge® Ultracentrifuge Rotors, Beckman Coulter®
Supplier: Beckman Coulter
Airfuge®
347833 347594 347844 341260 347593 347844 347596 341260 347594 347833 347596 347595 347595 347593
BK347844EA 5485.69 USD InStock
CABK347596 BK347844 BK347833 CABK347595 CABK347594 CABK341260 CABK347593 BK347595 BK347596 BK347593 BK347594 CABK347833 CABK347844 BK341260
Airfuge® Ultracentrifuge Rotors, Beckman Coulter®
Centrifuges Rotors
These rotors are for use with the Airfuge® Ultracentrifuge. (See BK340400 series.) Choose from a variety of fixed-angle and special application rotors.
  • Anodized Base
  • Stainless Steel Cover
  • Opening on Top
The rotor features an anodized base machined to fully support both chambers of the plastic liner
The cover is screwed down on the rotor base, compressing the liner and sealing the outer chamber from the inner chamber.
It has a stainless steel cover with an opening on the top, through which protrudes the dome of the inner chamber with the sample access hole.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty