Would you like to log in again?
Tube Racks for Aerosolve® Canisters, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Aerosolve®
359483 359487 359160 359484 359488 359161 359489 359486 359163 358991 359164 358992 354495 359164 359162 356985 359487 359486 359482 358992 356985 359162 359485 359160 358993 358991 359485 359161 359489 359488 359483 358993 359482 359163 359484
BK359487PK 923.68 USD InStock
CABK359488 CABK358993 CABK359489 CABK359486 CABK358991 CABK359487 CABK358992 CABK359484 CABK359485 BK359487 BK358992 BK359486 BK358991 BK359489 BK358993 BK359488 BK359164 BK354495 BK359483 BK359482 BK359163 BK359485 BK359484 BK356985 CABK356985 BK359160 BK359162 BK359161 CABK359163 CABK359482 CABK359483 CABK359164 CABK359161 CABK359162 CABK359160
Tube Racks for Aerosolve® Canisters, Beckman Coulter®
Racks Tube Racks
Molded plastic racks support tubes in Aerosolve® canisters.
  • Open-Rack Design for Quick Inspection
  • Molded Plastic Racks
  • Variety of Colors
The open-rack design permits quick inspection for possible broken or leaking tubes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty