Would you like to log in again?
Modular Disk Adapters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter®
Supplier: Beckman Coulter
359470 359155 359478 359472 354511 359472 359149 359469 359469 359476 359470 359148 359154 359475 359155 354511 359473 359149 359153 359473 359477 359153 359476 359150 359475 359151 359156 359152 359157 359150 359154 359471 359474 359151 359157 359474 359478 359471 359152 359156 359148 359477
BK359469PK 414.55 USD InStock
CABK359149 CABK359469 CABK359148 BK359469 BK359149 BK354511 BK359148 CABK354511 BK359470 BK359151 BK359150 CABK359471 CABK359152 CABK359472 CABK359153 CABK359150 CABK359470 CABK359151 CABK359477 CABK359478 CABK359475 CABK359156 CABK359476 CABK359157 CABK359473 CABK359154 CABK359155 CABK359474 BK359476 BK359157 BK359475 BK359156 BK359478 BK359477 BK359472 BK359153 BK359152 BK359471 BK359155 BK359474 BK359154 BK359473
Modular Disk Adapters for SX4750, SX4750A, and JS-4.3 Rotors, Beckman Coulter®
Centrifuges Rotors
Adapters are for use with the SX4750 series, GH3.8, and JS-4.3 (BK362734) rotors.
  • Durable Modular Units
  • Thick Rubber Base
  • Autoclavable
  • Alternative Application: Storage Rack
For spinning 1.5 ml microcentrifuge tubes, a 26-hole adapter plate fits over the 12 mm blue adapters (BK359148) and (BK359469).
Durable modular units stack onto a thick rubber base to accommodate a wide variety of laboratory tubes. All units are autoclavable and can also be used as storage racks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty