Would you like to log in again?
JS-24.15 and JS-24.38 Swinging Bucket Rotors, Titanium Buckets, Beckman Coulter®
Supplier: Beckman Coulter
362396 362396 360743 360743
BK362396EA 20180.51 USD InStock
CABK360743 BK362396 BK360743 CABK362396
JS-24.15 and JS-24.38 Swinging Bucket Rotors, Titanium Buckets, Beckman Coulter®
Centrifuges Rotors
The six-place, aluminum rotors with titanium swinging buckets are for use in Avanti™ J-26 XP and XPI and J6-MI centrifuge.
  • Ideal for Harvesting Bacteria, Processing Tissue Homogenates, Separating Cell Particulates, and More
  • Space for Six 15 mL Tubes or Six 38.5 mL Tubes
  • Titanium Swinging Buckets
  • Material: Aluminum
  • Six-Place Rotors
They are ideal for harvesting bacteria and subcellular particulates, processing tissue homogenates, and routine pelleting such as precipitates and phase separations.
They accommodate six 15mL tubes or six 38.5mL tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty