Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

JA-12 Fixed-Angle Rotors, Beckman Coulter®

Supplier: Beckman Coulter

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

360992 360993 360993 360992
BK360993EA 9855.5 USD InStock
BK360993 BK360992 CABK360992 CABK360993
JA-12 Fixed-Angle Rotors, Beckman Coulter®
Centrifuges Rotors
Aluminum, 12-place rotors are for use in Avanti™ centrifuges.

  • Ideal for Pelleting Cells, Bacteria, or Subcellular Fractions
  • Rotor Material: Anodized Aluminum
  • Rotor Capacity: 600 mL
  • Maximum Speed: 12,000 rpm
  • Accommodate Twelve 50mL Conical Tubes

They are ideal for pelleting cells, bacteria, or subcellular fractions as well as for performing phase separations and binding studies.

The rotors accommodate twelve 50mL conical tubes.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty