Would you like to log in again?
Rotors for Allegra® X-30 Series Centrifuges, Swinging-Bucket Microplate, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B01430 B01430 361111 361111
BK361111EA 3835.27 USD InStock
CABKB01430 BK361111 CABK361111 BKB01430
Rotors for Allegra® X-30 Series Centrifuges, Swinging-Bucket Microplate, Beckman Coulter®
Centrifuges Rotors
These rugged Beckman Coulter rotors are constructed from heavy-duty aluminum and are engineered to provide quiet, dependable service.
  • Spins 2 x 3 microplates or 2 x 1 deep-well plates
  • Applications include serial dilution of small, liquid volumes
  • Unshielded
  • Anodized aluminum material
  • Quiet
Ordering information: Microplates not included. Rotors are supplied complete and ready to use. Bottles, wrenches, and accessories are available separately. Contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty