Would you like to log in again?
Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
392243 392274 392271 392274 361171 361171 392271
BK392271EA 2653.1 USD InStock
CABK392274 CABK361171 CABK392243 BK392271 CABK392271 BK392274 BK361171
Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Centrifuges Rotors
These rugged rotors are constructed from heavy-duty aluminum and are engineered to provide quiet, dependable service.
  • Application: Pelleting Subcellular Organelles, Nucleic acids, Viruses, Bacteria, and More
  • Material: Heavy-Duty Aluminum
  • Quiet Process
  • Bio-Certified
  • Fixed, 45⁰ Angle
Fixed-angle rotors run at higher speeds for small volumes; swinging bucket rotors process large volumes at slower speeds.
Rotors accommodate tubes and bottles from 1.5 to 250 mL, up to six standard microplates, or two deep-well plates with a 96-well format. Rotors can be autoclaved or chemically sterilized.
Ordering information: Rotors are supplied complete and ready to use. Bottles and tubes are available separately. For bottle and tube adapters, see BK392261, BK361247, and BK392270 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty