Would you like to log in again?
JA-25.15 Fixed-Angle Rotors, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
363050 363054 363054 363050
BK363054EA 10403.68 USD InStock
BK363054 CABK363050 CABK363054 BK363050
JA-25.15 Fixed-Angle Rotors, Beckman Coulter®
Centrifuges Rotors
These aluminum, 24-place, and fixed-angle rotors are for use in Avanti™ J-26 XP, XPI, and J6-MI centrifuges.
  • Applications: Harvesting Bacteria, Routine Pelleting, and More
  • Material: Aluminum
  • Fixed-Angle
  • Space for Twenty-Four 15 mL Tubes
  • Biosafety Lid
They are ideal for harvesting bacteria, processing tissue homogenates and subcellular particulates, routine pelleting such as precipitates, and phase separation.
Rotors accommodate twenty-four 15 mL tubes and feature a Biosafety lid.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty