Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

J-Lite™ JLA-16.250 Fixed-Angle Rotors, Beckman Coulter®

Supplier: Beckman Coulter
J-Lite™
363930 363934 363930 363934
BK363930EA 12319.61 USD InStock
CABK363934 BK363930 BK363934 CABK363930
J-Lite™ JLA-16.250 Fixed-Angle Rotors, Beckman Coulter®
Centrifuges Rotors
These six-place, aluminum rotors are for use in Avanti™ J-26 XP, XPI, J-E, and J6-MI centrifuges.

  • Ideal for General Purpose, Large-Volume, and Multi-Tube Processing
  • Six-Place Rotors
  • Material: Aluminum
  • Weight: 10.3 kg
  • Space for Six 250 mL Tubes

At only 10.3 kg (22 7/10 lbs.), the rotors are a lightweight alternative to conventional 6 x 250 mL rotors

They are ideal for general purpose, large-volume, and multi-tube processing.

Rotors accommodate six 250 mL tubes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty