Would you like to log in again?
JS-4.2SM and JS-4.2SMA Swinging Bucket Rotors, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
348394 366670 348394 366670
BK348394EA 13962.03 USD InStock
CABK366670 CABK348394 BK348394 BK366670
JS-4.2SM and JS-4.2SMA Swinging Bucket Rotors, Beckman Coulter®
Centrifuges Rotors
Windshielded, six-place swinging bucket rotors feature oval-shaped buckets designed specifically for blood bank applications.
  • ARIES™ Smart Balance Technology Included
  • Specific Use for Blood Bank Applications
  • Six-Place, Windshielded Swinging Bucket Rotors
  • Space for Six Blood Bags
  • Imbalance Compensation for Buckets Unbalanced by up to 100 g.
Rotors JS-4.2SMA feature ARIES™ Smart Balance technology, which provides imbalance compensation for rotors with buckets that are unbalanced by up to 100 g.
These rotor are for use in the Avanti™ J6-MI centrifuge. Rotors accommodate six blood bags. The buckets are interchangeable with microplate carriers (sold separately); each carrier holds up to three microtiter plates.
Ordering information: Rotors include six oval-shaped blood bag cups.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty