Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TA-10-250 Fixed-Angle Rotor, Beckman Coulter®

Supplier: Beckman Coulter
368293 368293
BK368293EA 6545.4 USD InStock
CABK368293 BK368293
TA-10-250 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
This router is for use with the Allegra® 25R centrifuge.

  • Rapid Bacteria Pelleting up to 1.5 L
  • Flat-Bottom Bottles: 6 x 250 mL
  • Tubes: 6 x 50 mL
  • Maximum Speed: 10,000 rpm
  • Material: Aluminum

It's ideal for pelleting cells from large volumes or cell particles from tissue homogenates

The TA-10-250 rotor provides high volume and g force for applications where 15,300 g is required for pelleting.

Short-column methods, such as partially filled tubes, may also be used to purify large quantities of viruses in a cushion gradient.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty