Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TA-15-1.5 Fixed-Angle Rotor, Beckman Coulter®

Supplier: Beckman Coulter
368298 368298
BK368298EA 3295.51 USD InStock
CABK368298 BK368298
TA-15-1.5 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
This rotor is for use with the Allegra® 25R centrifuge.

  • Applications: DNA Precipitation, RNA Isolation, and Bacterial Call Pelleting for Mini-Preps
  • Maximum Speed: 15,000 rpm
  • Material: Aluminum
  • Imbalance Tolerance: 6 g
  • Nominal Rotor Capacity

It's ideal for DNA precipitation, RNA isolation, and bacterial cell pelleting for mini-preps

It accepts thirty 30 x 1.5-2.2 mL tubes or thirty 30-250 to 750 µL PCR tubes (requires adapter BK361247).

The rotor provides 24,700 g for sample volumes.

Ordering information: Rotor supplied with lid and O-ring.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty