Would you like to log in again?
Accessories for TS-5.1-500 Swinging Bucket Rotor, Beckman Coulter
Supplier: Beckman Coulter
368451 368451
BK368451PK 2115.96 USD InStock
CABK368451 BK368451
Accessories for TS-5.1-500 Swinging Bucket Rotor, Beckman Coulter
Centrifuges Rotors
This rotor is for use with the TS-5.1-500 rotor (BK368308) for the Allegra® 25R centrifuge (BK369434).
  • Applications: Cell Culture and Immunoassay Techniques, DNA Precipitation, Sequencing, and Cytocentrifugation
  • Holding Capacity: Up to Two Stacked 96-Well Microplates or One Deep-Well Plate
  • For Use in Benchtop Centrifuge Rotors
  • Rotor Provides 3010 g (4100 rpm) for Cell Culture and Immunoassay Technique Applications
Each microplate carrier can hold up to two stacked 96-well microplates or one deep-well plate.
It provides up to 3010 g (4100 rpm) for applications in cell culture and immunoassay techniques. It's also ideal for DNA precipitation, sequencing, and cytocentrifugation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty