Would you like to log in again?
JA-18 Fixed-Angle Rotor, Beckman Coulter®
Supplier: Beckman Coulter
369679 369679
BK369679EA 9268.55 USD InStock
BK369679 CABK369679
JA-18 Fixed-Angle Rotor, Beckman Coulter®
Centrifuges Rotors
This ten-place, aluminum, and fixed-angle rotor is for use in Avanti™ J-26 XP, XPI, J-E, and J6-MI centrifuges.
  • Application: Pelleting Bacteria, Cell Membranes, and Subcellular Organelles
  • Rotor Accommodates Ten 100 mL Tubes
  • Ten-Place
  • Material: Aluminum
  • Fixed-Angle
It is ideal for high-force, high-volume applications such as pelleting bacteria, cell membranes, and subcellular organelles.
The rotor accommodates ten 100 mL tubes.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty