Would you like to log in again?
Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
392243 392274 392271 392274 361171 361171 392271
BK392271EA 2653.1 USD InStock
CABK392274 CABK361171 CABK392243 BK392271 CABK392271 BK392274 BK361171
Rotors for Allegra® X-22 and Allegra X-22R Centrifuges, Beckman Coulter®
Centrifuges Rotors
These rugged rotors are constructed from heavy-duty aluminum and are engineered to provide quiet, dependable service.
  • Application: Pelleting Subcellular Organelles, Nucleic acids, Viruses, Bacteria, and More
  • Material: Heavy-Duty Aluminum
  • Quiet Process
  • Bio-Certified
  • Fixed, 45⁰ Angle
Fixed-angle rotors run at higher speeds for small volumes; swinging bucket rotors process large volumes at slower speeds.
Rotors accommodate tubes and bottles from 1.5 to 250 mL, up to six standard microplates, or two deep-well plates with a 96-well format. Rotors can be autoclaved or chemically sterilized.
Ordering information: Rotors are supplied complete and ready to use. Bottles and tubes are available separately. For bottle and tube adapters, see BK392261, BK361247, and BK392270 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty