Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories for Rotors, Beckman Coulter

Supplier: Beckman Coulter
392873 392806 393070 392805 392805 392873 393070 392806
BK392873PK 305.67 USD InStock
CABK392806 CABK392805 BK392873 CABK392873 BK393070 BK392805 BK392806 CABK393070
Accessories for Rotors, Beckman Coulter
Centrifuges Rotors
This assembly is compatible with Allegra® X-12, X-14, and X-15R series centrifuges (BK392472, BKA99464, and BK392932 series).

  • Application: Rapid Sediment of Protein Precipitates, Large Particles, Cells and Cell Debris
  • Material: Aluminum and Stainless Steel
  • Maximum Speed: 4450 rpm
  • Rubber Pads and Carriers Included

Each rotor has carriers that accommodate up to three standard microplates: one Beckman Coulter deep-well plate, one Beckman Coulter square-well plate, or one 96-tube rack.

It provides g forces up to 2890 g (3750 rpm) in Allegra X-12 and up to 4060 g (4450 rpm) in Allegra X-15R centrifuges for cellular assays and quick microplate spins.

Certifications: BioCertified covers are certified by CAMR, Porton Down, UK for biocontainment.

Ordering information: Each microplate carrier assembly includes two carriages and two rubber pads.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty