Would you like to log in again?
Allegra® X-30R Series Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Allegra®
B05796 B08540 B08708 B08536 B06314 B08542 B06314 B08547 B08538 B08545 B08542 B05802 B06320 B08705 B06320 B08545 B08536 B08540 B08547 B08705 B08708 B05796 B05802 B08538
BKB08708EA 31499.49 USD InStock
BKB08708 CABKB08705 CABKB05796 CABKB06314 BKB05802 BKB08705 CABKB08708 BKB08538 CABKB06320 BKB08536 CABKB08542 BKB08547 BKB05796 BKB06314 BKB08545 CABKB08545 CABKB05802 BKB06320 BKB08542 CABKB08536 CABKB08547 BKB08540 CABKB08538 CABKB08540
Allegra® X-30R Series Benchtop Centrifuges, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra® X-30R series benchtop centrifuges offer high performance and versatility in a compact design.
  • Cell Culture Package includes centrifuge and rotor SX4400
  • Microplate Application Package includes centrifuge and microplate rotor S606
  • Molecular Performance Application Package includes centrifuge and fixed-angle rotor F2402H
  • Clinical Applications Package includes centrifuge, rotor SX4400, and adapters
The Allegra® X-30R refrigerated models offer a g-force max of 29,756xg (18,000rpm) for fixed angle rotors and 4,255xg (4700rpm) for swing bucket rotors
Digital display indicates the actual speed, set speed, temperature (refrigerated model only), rotor imbalance, and ten different acceleration and deceleration rates. Four application packages are available for streamlined centrifuge solutions.
The Allegra® X-30R cools from room temperature to 4 °C in less than ten minutes. The refrigerated models have an operating temperature range of 2 to 40 °C (35 to 104°F).
The Allegra® X-30 ambient (non-refrigerated) models offer a g-force max of 23,511xg (16,000rpm) for fixed angle rotors and 4255xg (4700 rpm) for swing bucket rotors.
All models in this series accommodate samples from 0.25 to 400 ml tubes/bottles and standard 96-well micro and deep well plates. Each model features a brushless induction drive motor.
Certifications: UL and C-UL listed.
Ordering information: Rotors and accessories are also sold separately. Additional adapters are available. For more information, contact your Avantor representative.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty