Would you like to log in again?
Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
Avanti™
C19409 C19399 C19405 C19411 C19399 C19421 C19423 C19400 C19418 C01994 C19406 C19422 C19393 C19417 C19424 C19410 B99516 C19404 C01994 C19391 C19394 C19400 C19418 C19397 B99517 C19415 C19405 C19398 C19416 C19391 C19404 C19412 C19424 C19392 C19403 C19397 C19417 C19415 C19421 C19403 C19393 C19392 C19409 C19423 C19411 C19422 C19398 C01995 C19416 C19406 C19410 C19412 B99517 B99516 C01995 C19394
BKC19400EA 14768.41 USD InStock
CA76121-338 CA76121-336 CA76121-334 CABKC19412 CABKC19411 CABKC19418 CABKC19417 CABKC19416 CABKC19415 BKC19400 CABKC19410 BKC19404 BKC19405 BKC19403 BKC19409 BKC19406 76102-920 76102-924 CABKC19409 76102-922 CABKC19403 BKC19392 BKC19393 BKC19391 CABKC19400 CABKC19406 BKC19397 CABKC19405 BKC19394 CABKC19404 BKC19398 BKC19399 CA76121-314 BKC19422 BKC19423 CABKC19394 CABKC19393 CABKC19392 BKC19421 CABKC19399 CABKC19398 BKC19424 CABKC19397 CABKC19391 76102-904 CABKC19424 CABKC19423 CABKC19422 BKC19411 BKC19412 BKC19410 CABKC19421 BKC19415 BKC19416 BKC19417 BKC19418
Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Centrifuges Benchtop Centrifuges
The Avanti™ J-15 series of benchtop centrifuges (refrigerated or ventilated) leverage the ultra harmonic technology, designed to protect your sample and increase workflow time efficiencies.
  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance
The Avanti™ J-15 series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance.
Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), these centrifuges use ultra harmonic technology. Ultra harmonic technology leverages intelligent acceleration and deceleration profiles, reduces sample disturbances and provides time savings.
The Avanti™ J-15 series is an all around solution for your scientific needs: cell culture, transfection/transduction, nucleic acid purification, blood processing, assay set-up and more. Routine cell culture procedures are made easy and safe with the use of aerosolve canister. These optional BioCertified aerosolve canisters are made of clear shell to allow sample visibility even before the canister is opened.
Speed efficiency and minimum sample disturbances with ultra harmonic technology (intelligent acceleration and deceleration profiles).
Sample protection and higher yield: soft pellet deceleration profile protects the sample and results in increased sample yield. Optimize bench space (with our truly ventilated unit while processing any protocol in cell culture, blood separation or microbiology). Ease of use and faster accessibility: icon usability and intuitive display on high contrast color LCD screen.
Efficiency: Easy one-touch access to six programs and store up to 99 programs. JS-4.750 swinging bucket rotor. (Spin from 1.5 ml reaction vials to 750 ml bottles, single to quad-pack blood bags and multi-well plates, with the proper adapters or microplate carriers.)
JA-10.100 (6×100 ml) BioCertified fixed angle rotor. (Spin up to six 100 ml tubes at a 25° angle to the axis of rotation. The dual- locking lid allows the rotor to be removed from the centrifuge with the lid on.)
Applications for the JA-10.100 rotor include: density gradient separations of erythrocytes, cell lysate fractions, granules, as well as differential separation of DNA, proteins, and viruses. Up to 600 ml of sample volume can be centrifuged per run. Optional adapters allow spinning 1.5 µl microcentrifuge tubes, 10 ml round, 15 ml round and conical and 50 ml round and conical tubes.
BioCertified: is a term used to describe our products which have been tested and validated to demonstrate containment of microbiological aerosols by an independent, third-party facility. Improper use or maintenance may affect seal integrity and , this, containment. BioSafe and BioSafety are terms intended to describe the enhanced biocontainment features of our products.
Up to three plates can be run per carrier at maximum multi-well plate speeds. If running four plates, reduce run speed to 2700 rpm. Observe manufacturers run speed and stacking limitations if using other manufacturers multi-well plates.
Up to three plates, separated by cap strips, can be run per carrier at maximum multi-well plate speeds. If running four plates, cap strips cannot be used due to height limitations; reduce run speed to 2700 rpm. Observe manufacturers run speed and stacking limitations if using other manufacturers multi-well plates.
Certifications: Meets ASTM standards. EN-342.
Ordering information: The centrifuge is supplied with a one-year parts and labor warranty. Rotors, adapters and accessories are sold separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty