Would you like to log in again?
Avanti™ J-15 Series Benchtop Centrifuges, 200-230 V, Beckman Coulter®
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Avanti™
C19414 C38179 C19401 C38184 C38180 C19401 C38183 C38185 C19390 C19414 C38181 B99514 C19420 C38176 C19413 C38175 C38179 C38180 C19389 C38182 C38177 C31546 C31546 C19390 C38181 C19395 C19402 C19402 C38185 C19407 C19396 C19419 C38178 B99515 C19413 C31547 C19396 C38184 C38176 C38178 C38175 C38186 C19420 C19408 C19408 C38177 C19395 C31547 C19389 C19407 C38186 C38182 C19419 C38183
BKC19401EA 20669.38 USD InStock
CABKC19419 CABKC19414 CABKC19413 BKC19401 CA76332-698 CA76332-696 BKC19402 76332-088 BKC19408 BKC19407 CABKC19408 CABKC19402 CABKC19401 BKC19390 CA76332-758 BKC19396 CABKC19407 CA76332-756 BKC19395 CA76332-764 76332-114 CA76332-762 76332-112 CA76332-760 76332-110 CA76332-706 CA76332-704 CA76332-702 CABKC19395 CA76332-754 BKC19389 76332-108 CA76332-752 BKC19420 76332-106 76332-104 76332-102 CABKC19396 76332-100 CABKC19390 76332-092 76332-090 76102-906 76102-908 CABKC19389 CA76332-700 CA76332-740 CABKC19420 BKC19413 BKC19414 BKC19419 76332-098 76332-096 76332-094
Avanti™ J-15 Series Benchtop Centrifuges, 200-230 V, Beckman Coulter®
Centrifuges Benchtop Centrifuges
The Avanti™ J-15 series of benchtop centrifuges (refrigerated or ventilated) leverage the ultra harmonic technology, designed to protect your sample and increase workflow time efficiencies.
  • 200-230 V models, not for use in USA
  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance
The Avanti™ J-15 series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance.
Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), these centrifuges use ultra harmonic technology. Ultra harmonic technology leverages intelligent acceleration and deceleration profiles, reduces sample disturbances and provides time savings.
The Avanti™ J-15 series is an all around solution for your scientific needs: cell culture, transfection/transduction, nucleic acid purification, blood processing, assay set-up and more. Routine cell culture procedures are made easy and safe with the use of aerosolve canister. These optional BioCertified aerosolve canisters are made of clear shell to allow sample visibility even before the canister is opened.
Speed efficiency and minimum sample disturbances with ultra harmonic technology (intelligent acceleration and deceleration profiles).
Sample protection and higher yield: soft pellet deceleration profile protects the sample and results in increased sample yield. Optimize bench space (with our truly ventilated unit while processing any protocol in cell culture, blood separation or microbiology). Ease of use and faster accessibility: icon usability and intuitive display on high contrast color LCD screen.
Efficiency: Easy one-touch access to six programs and store up to 99 programs. JS-4.750 swinging bucket rotor. (Spin from 1.5 ml reaction vials to 750 ml bottles, single to quad-pack blood bags and multi-well plates, with the proper adapters or microplate carriers.)
JA-10.100 (6×100 ml) BioCertified fixed angle rotor. (Spin up to six 100 ml tubes at a 25° angle to the axis of rotation. The dual- locking lid allows the rotor to be removed from the centrifuge with the lid on.)
Applications for the JA-10.100 rotor include: density gradient separations of erythrocytes, cell lysate fractions, granules, as well as differential separation of DNA, proteins, and viruses. Up to 600 ml of sample volume can be centrifuged per run. Optional adapters allow spinning 1.5 µl microcentrifuge tubes, 10 ml round, 15 ml round and conical and 50 ml round and conical tubes.
BioCertified: is a term used to describe our products which have been tested and validated to demonstrate containment of microbiological aerosols by an independent, third-party facility. Improper use or maintenance may affect seal integrity and , this, containment. BioSafe and BioSafety are terms intended to describe the enhanced biocontainment features of our products.
Up to three plates can be run per carrier at maximum multi-well plate speeds. If running four plates, reduce run speed to 2700 rpm. Observe manufacturers run speed and stacking limitations if using other manufacturers multi-well plates.
Up to three plates, separated by cap strips, can be run per carrier at maximum multi-well plate speeds. If running four plates, cap strips cannot be used due to height limitations; reduce run speed to 2700 rpm. Observe manufacturers run speed and stacking limitations if using other manufacturers multi-well plates.
Certifications: Meets ASTM standards. EN-342.
Ordering information: The centrifuge is supplied with a one-year parts and labor warranty. Rotors, adapters and accessories are sold separately.
Caution: 200-230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (76102-920, 76102-922, 76102-904 and 76102-924) are also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty